J-502380

Supplier: BACHEM AMERICAS INC

4019023.0001 4019023.0005
B-2310.0001BAEA 542.13 CAD
B-2310.0001BA B-2310.0005BA
J-502380
J-502380

Sequence: Fmoc-Glu(OSu)-OtBu


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₂₈H₃₀N₂O₈
Storage Temperature: Freezer
MDL Number: MFCD00237021
CAS Number: 200616-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR