Fmoc-d-asp-otbu

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-D-aspartic acid 1-tert-butyl ester, Fmoc-D-Asp-OtBu

4019769.0001 4019769.0005
B-2305.0001BAEA 541.99 CAD
B-2305.0001BA B-2305.0005BA
Fmoc-d-asp-otbu
Fmoc-d-asp-otbu

Sequence: Fmoc-D-Asp-OtBu


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₂₃H₂₅NO₆
MW: 411.454 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00237011
CAS Number: 134098-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR