Fmoc-Dab-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-(Fmoc-amino)-4-aminobutyric acid

4012085.0001 4012085.0005
B-2300.0001BAEA 258.09 CAD
B-2300.0001BA B-2300.0005BA
Fmoc-Dab-OH
Fmoc-Dab-OH

Sequence: Fmoc-Dab-OH

Formula: C₁₉H₂₀N₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00237017
CAS Number: 161420-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR