(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid
Supplier: BACHEM AMERICAS INC
Synonyms:
(S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-2-carboxylic acid, Fmoc-Pip-OH, 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-pipecolic Acid, Fmoc-HoPro-OH, 1-Fmoc-L-pipecolic Acid, Fmoc-HomoPro-OH, (S)-1-Fmoc-2-piperidinecarboxylic Acid
4019015.0001
4019015.0005
B-2285.0001BAEA
439.41
CAD
B-2285.0001BA
B-2285.0005BA
(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid
(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid
Sequence: Fmoc-Homopro-OH
Pip (or Hpr) derivatives.
Synonym(s): Fmoc-L-pipecolic acid#Fmoc-Pip-OH
Formula:
C₂₁H₂₁NO₄ MW: 351.40 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00235898 CAS Number: 86069-86-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...