(S)-2,5-Bis((((9H-fluoren-9-yl)methoxy)carbonyl)amino)pentanoic acid

Supplier: BACHEM AMERICAS INC

4018519.0001 4018519.0005 4018519.0025
B-2260.0001BAEA 180.91 CAD
B-2260.0001BA B-2260.0005BA B-2260.0025BA
(S)-2,5-Bis((((9H-fluoren-9-yl)methoxy)carbonyl)amino)pentanoic acid
(S)-2,5-Bis((((9H-fluoren-9-yl)methoxy)carbonyl)amino)pentanoic acid

Sequence: Fmoc-Orn(Fmoc)-OH


Bachem also offers Fmoc-Orn(Boc)-Wang resin (D-1800).


Building block for the synthesis of MAP cores.

Formula: C₃₅H₃₂N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00153362
CAS Number: 201046-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR