Fmoc-4,5-dehydro-l-leucine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-4,5-dehydro-Leu-OH

4018956.0001 4018956.0005
B-2255.0001BAEA 1086.93 CAD
B-2255.0001BA B-2255.0005BA
Fmoc-4,5-dehydro-l-leucine
Fmoc-4,5-dehydro-l-leucine

Sequence: Fmoc-4,5-dehydro-Leu-OH

Formula: C₂₁H₂₁NO₄
MW: 351.402 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237651
CAS Number: 87720-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR