Fmoc-L-homocitrulline

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Homocit-OH, Fmoc-HoCit-OH, Nα-Fmoc-Nepsilon-carbamoyl-L-lysine

4019724.0001 4019724.0005
B-2250.0001BAEA 439.41 CAD
B-2250.0001BA B-2250.0005BA
Fmoc-L-homocitrulline
Fmoc-L-homocitrulline

Sequence: Fmoc-Homocit-OH


Hci derivatives.


Synonym(s): Fmoc-Lys(carbamoyl)-OH

Formula: C₂₂H₂₅N₃O₅
MW: 411.458 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00237387
CAS Number: 201485-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR