Fmoc-D-Glu-OtBu
Supplier: BACHEM AMERICAS INC
4018580.0001
4018580.0005
B-2245.0001BAEA
335.58
CAD
B-2245.0001BA
B-2245.0005BA
Fmoc-D-Glu-OtBu
Fmoc-D-Glu-OtBu
Sequence: Fmoc-D-Glu-OtBu
The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
Formula:
C₂₄H₂₇NO₆ Storage Temperature: Ambient |
MDL Number:
MFCD00077054 CAS Number: 109745-15-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...