Fmoc-Lys(Alloc)-OH

Supplier: BACHEM AMERICAS INC

4016656.0005 4016656.0025
B-2240.0005BAEA 233.1 CAD
B-2240.0005BA B-2240.0025BA
Fmoc-Lys(Alloc)-OH
Fmoc-Lys(Alloc)-OH

Sequence: Fmoc-Lys(Aloc)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".


The selectively cleavable Aloc group allows on-resin modification of lysine side-chains during Fmoc-SPPS.

Formula: C₂₅H₂₈N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00190872
CAS Number: 146982-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR