Fmoc-D-allo-isoleucine

Supplier: BACHEM AMERICAS INC

4018803.0001 4018803.0005
B-2230.0001BAEA 793.9 CAD
B-2230.0001BA B-2230.0005BA
Fmoc-D-allo-isoleucine
Fmoc-D-allo-isoleucine

Sequence: Fmoc-D-allo-Ile-OH

Formula: C₂₁H₂₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00077062
CAS Number: 118904-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR