Fmoc-Bpa-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-4-benzoyl-L-phenylalanine, Fmoc-p-Bz-Phe-OH

4017859.0001 4017859.0005
B-2220.0001BAEA 413.58 CAD
B-2220.0001BA B-2220.0005BA
Fmoc-Bpa-OH
Fmoc-Bpa-OH

Sequence: Fmoc-p-Bz-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.
4-Bpa is incorporated into peptides as a photolabel or for photocrosslinking.


Synonym(s): Fmoc-Bpa-OH

Formula: C₃₁H₂₅NO₅
MW: 491.543 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151937
CAS Number: 117666-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR