(S)-Perfluorophenyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-oxo-4-(tritylamino)butanoate

Supplier: BACHEM AMERICAS INC

4017844.0001 4017844.0005
B-2200.0001BAEA 175.66 CAD
B-2200.0001BA B-2200.0005BA
(S)-Perfluorophenyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-oxo-4-(tritylamino)butanoate
(S)-Perfluorophenyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-oxo-4-(tritylamino)butanoate

Sequence: Fmoc-Asn(Trt)-OPfp


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".

Formula: C₄₄H₃₁F₅N₂O₅
Storage Temperature: Freezer
MDL Number: MFCD00153373
CAS Number: 132388-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR