Fmoc-N-Me-Asp(OtBu)-OH

Supplier: BACHEM AMERICAS INC

4018551.0001 4018551.0005
B-2195.0001BAEA 849.55 CAD
B-2195.0001BA B-2195.0005BA
Fmoc-N-Me-Asp(OtBu)-OH
Fmoc-N-Me-Asp(OtBu)-OH

Sequence: Fmoc-N-Me-Asp(OtBu)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.
Bachem also offers the neurotransmitter NMDA, F-2145.

Formula: C₂₄H₂₇NO₆
MW: 425.481 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237027
CAS Number: 152548-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR