Fmoc-His(Mtt)-OH

Supplier: BACHEM AMERICAS INC

4018533.0005 4018533.0025
B-2190.0005BAEA 284.04 CAD
B-2190.0005BA B-2190.0025BA
Fmoc-His(Mtt)-OH
Fmoc-His(Mtt)-OH

Sequence: Fmoc-His(1-Mtt)-OH


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): Fmoc-His(τ-Mtt)-OH

Formula: C₄₁H₃₅N₃O₄
Storage Temperature: Freezer
MDL Number: MFCD00153358
CAS Number: 133367-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR