Fmoc-Leu-Pro-OH

Supplier: BACHEM AMERICAS INC

4017133.0001 4017133.0005
B-2185.0001BAEA 464.74 CAD
B-2185.0001BA B-2185.0005BA
Fmoc-Leu-Pro-OH
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/peptide name Fmoc-Leu-Pro-OH
Environmentally Preferable
Species
Source
CAS No. 130832-25-6
MDL No. MFCD00237660
Sequence Fmoc-Leu-Pro-OH
Molecular weight 450.53
Molecular formula C₂₆H₃₀N₂O₅
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR