Fmoc-Asp-ODmb

Supplier: BACHEM AMERICAS INC

4018284.0001 4018284.0005
B-2175.0001BAEA 672.04 CAD
B-2175.0001BA B-2175.0005BA
Fmoc-Asp-ODmb
Fmoc-Asp-ODmb

Sequence: Fmoc-Asp-ODmb


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.


The highly acid-labile Dmb ester can be cleaved, e.g., with 1%TFA/DCM.

Formula: C₂₈H₂₇NO₈
Storage Temperature: Freezer
MDL Number: MFCD00237652
CAS Number: 155866-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR