Fmoc-Met(O)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-L-methionine sulphoxide, N-Fmoc-L-methionine sulfoxide

4013494.0001 4013494.0005
B-2130.0001BAEA 180.91 CAD
B-2130.0001BA B-2130.0005BA
Fmoc-Met(O)-OH
Fmoc-Met(O)-OH

Sequence: Fmoc-Met(O)-OH


Oxidation of Met to the sulfoxide Met(O) can be easily reverted, whereas sulfone (Met(O₂)) formation is practically irreversible.


(Diastereomeric mixture)

Formula: C₂₀H₂₁NO₅S
MW: 387.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077064
CAS Number: 76265-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR