Fmoc-D-Trp(Boc)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
1-Boc-N-Fmoc-D-tryptophan
4017826.0001
4017826.0005
B-2125.0001BAEA
227.22
CAD
B-2125.0001BA
B-2125.0005BA
Fmoc-D-Trp(Boc)-OH
Fmoc-D-Trp(Boc)-OH
Sequence: Fmoc-D-Trp(Boc)-OH
Sold under license to US Pat. 5,300,651 and EP 0 516 070
Formula:
C₃₁H₃₀N₂O₆ MW: 526.589 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00153367 CAS Number: 163619-04-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...