Fmoc-4-chloro-L-phenylalanine

Supplier: BACHEM AMERICAS INC

4018280.0001 4018280.0005
B-2115.0001BAEA 309.81 CAD
B-2115.0001BA B-2115.0005BA
Fmoc-4-chloro-L-phenylalanine
Fmoc-4-chloro-L-phenylalanine

Sequence: Fmoc-4-chloro-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.


Synonym(s): Fmoc-p-chloro-Phe-OH#(S)-3-(4-Chloro-phenyl)-2-(Fmoc-amino)-propionic acid

Formula: C₂₄H₂₀ClNO₄
Storage Temperature: Ambient
MDL Number: MFCD00080281
CAS Number: 175453-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR