Fmoc-Tle-OH

Supplier: BACHEM AMERICAS INC

4016319.0001 4016319.0005 4016319.0025
B-2110.0001BAEA 387.21 CAD
B-2110.0001BA B-2110.0005BA B-2110.0025BA
Fmoc-Tle-OH
Fmoc-Tle-OH

Sequence: Fmoc-tBu-Gly-OH


A sterically demanding, lipophilic amino acid with a quaternary side-chain Cβ-atom. The bulky t-butyl group of Tle destabilizes α-helices.


Synonym(s): Fmoc-Tle-OH

Formula: C₂₁H₂₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00065649
CAS Number: 132684-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR