Fmoc-3-(2-naphthyl)-L-alanine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-2-Nal-OH

4017664.0001 4017664.0005 4017664.0025
B-2100.0001BAEA 387.21 CAD
B-2100.0001BA B-2100.0005BA B-2100.0025BA
Fmoc-3-(2-naphthyl)-L-alanine
Fmoc-3-(2-naphthyl)-L-alanine

Sequence: Fmoc-2-Nal-OH


Fluorescent phenylalanine analog.

Formula: C₂₈H₂₃NO₄
MW: 437.495 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144886
CAS Number: 112883-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR