(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-L-citrulline, Nalpha-Fmoc-L-citrulline, Fmoc-Cit-OH

4017699.0005 4017699.0025
B-2090.0005BAEA 309.81 CAD
B-2090.0005BA B-2090.0025BA
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid

Sequence: Fmoc-Cit-OH


Bachem also offers Fmoc-Cit-Wang resin, D-1840.


Synonym(s): Fmoc-Orn(carbamoyl)-OH

Formula: C₂₁H₂₃N₃O₅
Storage Temperature: Ambient
MDL Number: MFCD00151943
CAS Number: 133174-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR