Fmoc-D-Cit-OH

Supplier: BACHEM AMERICAS INC

4017700.0001 4017700.0005
B-2075.0001BAEA 387.21 CAD
B-2075.0001BA B-2075.0005BA
Fmoc-D-Cit-OH
Fmoc-D-Cit-OH

Sequence: Fmoc-D-Cit-OH


Bachem also offers Fmoc-Cit-Wang resin, D-1840.


Synonym(s): Fmoc-D-Orn(carbamoyl)-OH

Formula: C₂₁H₂₃N₃O₅
MW: 397.431 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151942
CAS Number: 200344-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR