Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N1-tert-butoxycarbonyl-L-tryptophan

Supplier: BACHEM AMERICAS INC

4017674.0005 4017674.0025 4017674.0100
B-2065.0005BAEA 175.66 CAD
B-2065.0005BA B-2065.0025BA B-2065.0100BA
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N1-tert-butoxycarbonyl-L-tryptophan
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N1-tert-butoxycarbonyl-L-tryptophan

Sequence: Fmoc-Trp(Boc)-OH


Sold under license to US Pat. 5,300,651 and EP 0 516 070. The use of this protected tryptophan derivative in Fmoc solid-phase peptide synthesis resulted in an improved yield of the correct product by suppressing side reactions of tryptophan.

Formula: C₃₁H₃₀N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00153366
CAS Number: 143824-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR