Fmoc-Gln(Mtt)-OH
Supplier: BACHEM AMERICAS INC
4017008.0001
4017008.0005
4017008.0025
B-2050.0001BAEA
175.66
CAD
B-2050.0001BA
B-2050.0005BA
B-2050.0025BA
Fmoc-Gln(Mtt)-OH
Fmoc-Gln(Mtt)-OH
Sequence: Fmoc-Gln(Mtt)-OH
For isoglutamine derivatives please see the family "Glutamic Acid".
Formula:
C₄₀H₃₆N₂O₅ Storage Temperature: Refrigerator |
MDL Number:
MFCD00237163 CAS Number: 144317-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...