Fmoc-3-(3-pyridyl)-D-alanine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-β-(3-pyridyl)-D-Ala-OH, Fmoc-D-3-Pal-OH

4017389.0001 4017389.0005
B-2040.0001BAEA 516.34 CAD
B-2040.0001BA B-2040.0005BA
Fmoc-3-(3-pyridyl)-D-alanine
Fmoc-3-(3-pyridyl)-D-alanine

Sequence: Fmoc-β-(3-pyridyl)-D-Ala-OH


3-Pal derivatives.


Fmoc-D-3-Pal-OH, building block for LHRH antagonists.

Formula: C₂₃H₂₀N₂O₄
MW: 388.423 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00080267
CAS Number: 142994-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR