Fmoc-N-methyl-L-leucine

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-N-methyl-L-leucine, Fmoc-N-Me-Leu-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-leucine

4017630.0001 4017630.0005
B-2035.0001BAEA 258.14 CAD
B-2035.0001BA B-2035.0005BA
Fmoc-N-methyl-L-leucine
Fmoc-N-methyl-L-leucine

Sequence: Fmoc-N-Me-Leu-OH


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).

Formula: C₂₂H₂₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD00151933
CAS Number: 103478-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR