N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine

Supplier: BACHEM AMERICAS INC

Synonyms: Nα-Fmoc-S-trityl-D-cysteine, Fmoc-D-Cys(Trt)-OH, N-Fmoc-S-trityl-D-cysteine

4017507.0001 4017507.0005 4017507.0025
B-2030.0001BAEA 175.66 CAD
B-2030.0001BA B-2030.0005BA B-2030.0025BA
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine

Sequence: Fmoc-D-Cys(Trt)-OH


Besides "standard" preloaded resins, Bachem offers laterally anchored Fmoc-cysteine, Fmoc-Cys(SASRIN resin)-OH (D-2170) and resin-linked Fmoc-cysteamine, Fmoc-cysteamine-SASRIN resin (D-2165).

Formula: C₃₇H₃₁NO₄S
Storage Temperature: Ambient
MDL Number: MFCD00151922
CAS Number: 167015-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR