N-Fmoc-N'-trityl-D-asparagine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-D-Asn(Trt)-OH

4017373.0001 4017373.0005 4017373.0025
B-2010.0001BAEA 201.47 CAD
B-2010.0001BA B-2010.0005BA B-2010.0025BA
N-Fmoc-N'-trityl-D-asparagine
N-Fmoc-N'-trityl-D-asparagine

Sequence: Fmoc-D-Asn(Trt)-OH


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".

Formula: C₃₈H₃₂N₂O₅
MW: 596.683 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151919
CAS Number: 180570-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR