Fmoc-His(Boc)-OPfp
Supplier: BACHEM AMERICAS INC
4015229.0001
4015229.0025
B-2000.0001BAEA
232.35
CAD
B-2000.0001BA
B-2000.0025BA
Fmoc-His(Boc)-OPfp
Fmoc-His(Boc)-OPfp
Sequence: Fmoc-His(Boc)-OPfp
The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.
Formula:
C₃₂H₂₆F₅N₃O₆ Storage Temperature: Freezer |
MDL Number:
MFCD00237049 CAS Number: 109053-20-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...