Fmoc-4-(boc-amino)-L-phenylalanine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Phe(4-NHBoc)-OH, Fmoc-4-(Boc-amino)-L-phenylalanine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-((tert-butoxycarbonyl)amino)phenyl)propanoic acid

4016065.0001 4016065.0005
B-1995.0001BAEA 413.58 CAD
B-1995.0001BA B-1995.0005BA
Fmoc-4-(boc-amino)-L-phenylalanine
Fmoc-4-(boc-amino)-L-phenylalanine

Sequence: Fmoc-p-amino-Phe(Boc)-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.
4-Aph derivatives.


Educt for the synthesis of p-guanidinyl-Phe derivatives.

Formula: C₂₉H₃₀N₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00237007
CAS Number: 174132-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR