Fmoc-Tyr(PO3Me2)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Tyr(PO₃Me₂)-OH

4017013.0001 4017013.0005
B-1990.0001BAEA 516.34 CAD
B-1990.0001BA B-1990.0005BA
Fmoc-Tyr(PO3Me2)-OH
Fmoc-Tyr(PO3Me2)-OH

Sequence: Fmoc-Tyr(PO₃Me₂)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
For stable phosphotyrosine mimics see the O-malonyltyrosine derivative B-2825 and the 4-phosphonophenylalanine derivatives B-3895 and A-4725.

Formula: C₂₆H₂₆NO₈P
Storage Temperature: Freezer
MDL Number: MFCD00133599
CAS Number: 127633-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR