N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine

Supplier: BACHEM AMERICAS INC

4018171.0001 4018171.0005 4018171.0025
B-1975.0001BAEA 206.51 CAD
B-1975.0001BA B-1975.0005BA B-1975.0025BA
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine

Sequence: Fmoc-β-cyclohexyl-Ala-OH


Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines
Cha corresponds to hydrogenated phenylalanine, the aromatic nucleus is replaced by a more flexible aliphatic cycle.


Synonym(s): Fmoc-Cha-OH

Formula: C₂₄H₂₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00065614
CAS Number: 135673-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR