Fmoc-3-(1-naphthyl)-L-alanine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-1-Nal-OH

4016166.0001 4016166.0005
B-1965.0001BAEA 476.62 CAD
B-1965.0001BA B-1965.0005BA
Fmoc-3-(1-naphthyl)-L-alanine
Fmoc-3-(1-naphthyl)-L-alanine

Sequence: Fmoc-1-Nal-OH


1-Nal and 2-Nal have been used as Trp replacements.

Formula: C₂₈H₂₃NO₄
MW: 437.495 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151914
CAS Number: 96402-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR