Fmoc-3-(2-naphthyl)-D-alanine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-D-2-Nal-OH

4017071.0001 4017071.0005 4017071.0025
B-1950.0001BAEA 376.89 CAD
B-1950.0001BA B-1950.0005BA B-1950.0025BA
Fmoc-3-(2-naphthyl)-D-alanine
Fmoc-3-(2-naphthyl)-D-alanine

Sequence: Fmoc-D-2-Nal-OH


Fluorescent phenylalanine analog.

Formula: C₂₈H₂₃NO₄
MW: 437.495 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077044
CAS Number: 138774-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR