Fmoc-L-norvaline

Supplier: BACHEM AMERICAS INC

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-norvaline, Fmoc-Nva-OH, N-Fmoc-L-norvaline

4016412.0005 4016412.0025
B-1945.0005BAEA 185.46 CAD
B-1945.0005BA B-1945.0025BA
Fmoc-L-norvaline
Fmoc-L-norvaline

Sequence: Fmoc-Nva-OH
Synonym(s): Fmoc-L-2-aminovaleric acid

Formula: C₂₀H₂₁NO₄
MW: 339.391 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00155631
CAS Number: 135112-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR