Fmoc-D-Tic-OH

Supplier: BACHEM AMERICAS INC

4015638.0001 4015638.0005
B-1925.0001BAEA 529.25 CAD
B-1925.0001BA B-1925.0005BA
Fmoc-D-Tic-OH
Fmoc-D-Tic-OH

Sequence: Fmoc-D-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid


Tic represents a constrained Phe analog.


Synonym(s): Fmoc-D-Tic-OH

Formula: C₂₅H₂₁NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00144345
CAS Number: 130309-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR