Fmoc-Tic-OH

Supplier: BACHEM AMERICAS INC

4015637.0001 4015637.0005
B-1920.0001BAEA 361.41 CAD
B-1920.0001BA B-1920.0005BA
Fmoc-Tic-OH
Fmoc-Tic-OH

Sequence: Fmoc-L-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid


Tic represents a constrained Phe analog.


Synonym(s): Fmoc-Tic-OH

Formula: C₂₅H₂₁NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00144368
CAS Number: 136030-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR