Fmoc-GABA-OH

Supplier: BACHEM AMERICAS INC

4016030.0001 4016030.0005 4016030.0025
B-1910.0001BAEA 180.91 CAD
B-1910.0001BA B-1910.0005BA B-1910.0025BA
Fmoc-GABA-OH
Fmoc-GABA-OH

Sequence: Fmoc-γ-Abu-OH


Flexible spacer molecules of various length.


Synonym(s): Fmoc-GABA-OH

Formula: C₁₉H₁₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD00144889
CAS Number: 116821-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR