Fmoc-Met(O2)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-L-methionine sulfone, Fmoc-Met(O₂)-OH, N-Fmoc-L-methionine sulphone

4013865.0001 4013865.0005
B-1905.0001BAEA 185.3 CAD
B-1905.0001BA B-1905.0005BA
Fmoc-Met(O2)-OH
Fmoc-Met(O2)-OH

Sequence: Fmoc-Met(O₂)-OH


Oxidation of Met to the sulfoxide Met(O) can be easily reverted, whereas sulfone (Met(O₂)) formation is practically irreversible.

Formula: C₂₀H₂₁NO₆S
Storage Temperature: Ambient
MDL Number: MFCD00153361
CAS Number: 163437-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR