Fmoc-4-chloro-D-phenylalanine
Supplier: BACHEM AMERICAS INC
Synonyms:
N-Fmoc-4-chloro-D-phenylalanine, Fmoc-D-Phe(4-Cl)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-chlorophenyl)propanoic acid, Fmoc-p-chloro-D-Phe-OH
Sequence: Fmoc-p-chloro-D-Phe-OH
Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.
Synonym(s): (R)-3-(4-Chloro-phenyl)-2-(Fmoc-amino)-propionic acid#Fmoc-4-chloro-D-Phe-OH
Formula:
C₂₄H₂₀ClNO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00077066 CAS Number: 142994-19-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...