Fmoc-D-Cys(Acm)-OH

Supplier: BACHEM AMERICAS INC

4014984.0001 4014984.0005
B-1890.0001BAEA 201.47 CAD
B-1890.0001BA B-1890.0005BA
Fmoc-D-Cys(Acm)-OH
Fmoc-D-Cys(Acm)-OH

Sequence: Fmoc-D-Cys(Acm)-OH


Besides "standard" preloaded resins, Bachem offers laterally anchored Fmoc-cysteine, Fmoc-Cys(SASRIN resin)-OH (D-2170) and resin-linked Fmoc-cysteamine, Fmoc-cysteamine-SASRIN resin (D-2165).

Formula: C₂₁H₂₂N₂O₅S
MW: 414.476 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077053
CAS Number: 168300-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR