Fmoc-Pen(Acm)-OH

Supplier: BACHEM AMERICAS INC

4016442.0001 4016442.0005
B-1885.0001BAEA 516.34 CAD
B-1885.0001BA B-1885.0005BA
Fmoc-Pen(Acm)-OH
Fmoc-Pen(Acm)-OH

Sequence: Fmoc-Pen(Acm)-OH


Corresponds to β,β-dimethylcysteine or β-mercaptovaline. For an S-protected derivative of deamino-penicillamine please see Q-1610. The rigidity of disulfide bridges can be increased by replacing Cys by Pen. Pen analogs of enkephalins, RGD-peptides, urotensin and further active peptides with improved potency and/or selectivity have been described. Bachem offers a choice of resins preloaded with Fmoc-Pen derivatives, please see the product family 'Preloaded Resins for SPPS and SPOS'.


Synonym(s): Fmoc-β,β-dimethyl-Cys(Acm)-OH

Formula: C₂₃H₂₆N₂O₅S
Storage Temperature: Ambient
MDL Number: MFCD00151935
CAS Number: 201531-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR