H-Lys(Fmoc)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
Nepsilon-Fmoc-L-lysine
4014126.0001
4014126.0005
4014126.0025
B-1875.0001BAEA
185.46
CAD
B-1875.0001BA
B-1875.0005BA
B-1875.0025BA
H-Lys(Fmoc)-OH
H-Lys(Fmoc)-OH
Sequence: H-Lys(Fmoc)-OH
For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
Formula:
C₂₁H₂₄N₂O₄ MW: 368.433 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00038365 CAS Number: 84624-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...