Fmoc-α-methylalanine
Supplier: BACHEM AMERICAS INC
Synonyms:
N-Fmoc-alpha-methylalanine, 2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]isobutyric acid, Fmoc-Aib-OH, 2-(Fmoc-amino)isobutyric acid
4013932.0005
4013932.0025
4013932.0100
B-1860.0005BAEA
335.58
CAD
B-1860.0005BA
B-1860.0025BA
B-1860.0100BA
Fmoc-α-methylalanine
Fmoc-alpha-methylalanine
Sequence: Fmoc-Aib-OH
Synonym(s): Fmoc-α-Me-Ala-OH
The acid fluoride of this sterically hindered amino acid could be coupled to N-methylated amino acids in HFIP at 55°C.
Formula:
C₁₉H₁₉NO₄ MW: 325.364 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00151913 CAS Number: 94744-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...