Fmoc-His(Boc)-OH

Supplier: BACHEM AMERICAS INC

4010990.0005 4010990.0025 4010990.0100
B-1830.0005BAEA 321.59 CAD
B-1830.0005BA B-1830.0025BA B-1830.0100BA
Fmoc-His(Boc)-OH
Fmoc-His(Boc)-OH

Sequence: Fmoc-His(1-Boc)-OH


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): Fmoc-His(τ-Boc)-OH

Formula: C₂₆H₂₇N₃O₆
Storage Temperature: Freezer
MDL Number: MFCD00151928
CAS Number: 81379-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR