Fmoc-D-His(Boc)-OH

Supplier: BACHEM AMERICAS INC

4014156.0001 4014156.0005 4014156.0025
B-1825.0001BAEA 284.04 CAD
B-1825.0001BA B-1825.0005BA B-1825.0025BA
Fmoc-D-His(Boc)-OH
Fmoc-D-His(Boc)-OH

Sequence: Fmoc-D-His(Boc)-OH


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.

Formula: C₂₆H₂₇N₃O₆
Storage Temperature: Freezer
MDL Number: MFCD00151927
CAS Number: 159631-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR