Fmoc-allo-Thr(tBu)-OH

Supplier: BACHEM AMERICAS INC

4016123.0001 4016123.0005
B-1815.0001BAEA 672.04 CAD
B-1815.0001BA B-1815.0005BA
Fmoc-allo-Thr(tBu)-OH
Fmoc-allo-Thr(tBu)-OH

Sequence: Fmoc-allo-Thr(tBu)-OH


Bachem also offers phosphothreonine derivatives.
aThr derivatives.

Formula: C₂₃H₂₇NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD00237371
CAS Number: 201481-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR