Fmoc-Dab(Boc)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (2S)-2,4-Diaminobutanoic acid, 4-BOC, 2-FMOC protected

4016009.0001 4016009.0005
B-1800.0001BAEA 370.68 CAD
B-1800.0001BA B-1800.0005BA
Fmoc-Dab(Boc)-OH
Fmoc-Dab(Boc)-OH

Sequence: Fmoc-Dab(Boc)-OH


Used in combination with the Nγ-Z derivative for the synthesis of polymyxin analogs.

Formula: C₂₄H₂₈N₂O₆
MW: 440.496 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00151941
CAS Number: 125238-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR