Nα-Fmoc-N-δ-trityl-L-glutamine

Supplier: BACHEM AMERICAS INC

4014949.0005 4014949.0025 4014949.0100
B-1790.0005BAEA 175.66 CAD
B-1790.0005BA B-1790.0025BA B-1790.0100BA
Nα-Fmoc-N-δ-trityl-L-glutamine
N-alpha-Fmoc-N-delta-trityl-L-glutamine

Sequence: Fmoc-Gln(Trt)-OH


For isoglutamine derivatives please see the family "Glutamic Acid".

Formula: C₃₉H₃₄N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00077056
CAS Number: 132327-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR